Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmnl2
Entrez Gene ID
71409
Gene Name
formin-like 2
Gene Aliases
5430425K04Rik, Man
Chromosome Location
Chr.2: 52857716 - 53134202 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172409.2 NP_765997.2 10 - 11 1226 155
GenBank mRNA CA329111.1 - 6 - 7 597 155

View Full Product Details