The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145824.1 | NP_001139296.1 | 12 - 13 | 3142 | 97 |
NM_010434.2 | NP_034564.2 | 12 - 13 | 3145 | 97 | |
GenBank mRNA | AF077660.1 | - | 12 - 13 | 2899 | 97 |
AF170305.1 | - | 11 - 12 | 2609 | 97 | |
AF305238.1 | - | 11 - 12 | 2607 | 97 | |
AK140130.1 | - | 12 - 13 | 3248 | 97 | |
BC131663.1 | - | 12 - 13 | 2777 | 97 | |
BC131664.1 | - | 12 - 13 | 2773 | 97 |