The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025301.2 | NP_079577.1 | 2 - 3 | 347 | 71 |
GenBank mRNA | AK007438.1 | - | 1 - 2 | 72 | 71 |
AK007828.1 | - | 2 - 3 | 328 | 71 | |
AK012906.1 | - | 2 - 3 | 217 | 71 | |
AK088838.1 | - | 2 - 3 | 240 | 71 | |
AK144092.1 | - | 2 - 3 | 288 | 71 | |
AK147957.1 | - | 2 - 3 | 282 | 71 |