The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039645.1 | NP_001034734.1 | 3 - 4 | 311 | 65 |
GenBank mRNA | AK043553.1 | - | 3 - 4 | 506 | 65 |
AK134834.1 | - | 2 - 3 | 1002 | 65 | |
BC111101.1 | - | 3 - 4 | 311 | 65 |