Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcn1
Entrez Gene ID
19672
Gene Name
reticulocalbin 1
Gene Aliases
Rcn
Chromosome Location
Chr.2: 105385948 - 105399319 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009037.2 NP_033063.1 2 - 3 543 100
GenBank mRNA AK017494.1 - 2 - 3 450 100
AK133971.1 - 2 - 3 493 100
AK159973.1 - 2 - 3 446 100
BC049108.1 - 2 - 3 436 100
D13003.1 - 2 - 3 463 100

View Full Product Details