Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fpr-rs3
Entrez Gene ID
14290
Gene Name
formyl peptide receptor, related sequence 3
Gene Aliases
-
Chromosome Location
Chr.17: 20623846 - 20624877 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008040.2 NP_032066.2 1 - 1 994 63
GenBank mRNA BC134352.1 - 1 - 1 989 63

View Full Product Details