The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205216.1 | NP_001192145.1 | 7 - 8 | 1123 | 78 |
NM_011306.4 | NP_035436.1 | 7 - 8 | 1400 | 78 | |
GenBank mRNA | BC049773.1 | - | 7 - 8 | 1360 | 78 |
KJ663717.1 | - | 7 - 8 | 1214 | 78 | |
KJ663719.1 | - | 8 - 9 | 1100 | 78 | |
M26804.1 | - | 6 - 7 | 992 | 78 | |
M84818.1 | - | 7 - 8 | 1073 | 78 | |
X66224.1 | - | 6 - 7 | 992 | 78 |