Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim10
Entrez Gene ID
19824
Gene Name
tripartite motif-containing 10
Gene Aliases
AI324236, BB139825, Herf1, Rnf9
Chromosome Location
Chr.17: 36869574 - 36877833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011280.2 NP_035410.2 1 - 2 732 75
GenBank mRNA AF220121.1 - 1 - 2 719 75
AK011082.1 - 1 - 2 732 75
BC051632.1 - 1 - 2 705 75

View Full Product Details