The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011280.2 | NP_035410.2 | 6 - 7 | 1234 | 115 |
GenBank mRNA | AF134811.1 | - | 6 - 7 | 1216 | 115 |
AF220121.1 | - | 6 - 7 | 1221 | 115 | |
AK011082.1 | - | 6 - 7 | 1234 | 115 | |
BC051632.1 | - | 6 - 7 | 1207 | 115 |