The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018867.5 | NP_061355.3 | 8 - 9 | 1254 | 69 |
GenBank mRNA | AF017639.1 | - | 8 - 9 | 1255 | 69 |
AK019509.1 | - | 8 - 9 | 1253 | 69 | |
BC021444.1 | - | 8 - 9 | 1176 | 69 |