Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bub3
Entrez Gene ID
12237
Gene Name
budding uninhibited by benzimidazoles 3 homolog (S. cerevisiae)
Gene Aliases
AU019800, AU021329, AU043350, AW146323, C78067
Chromosome Location
Chr.7: 131560370 - 131571898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009774.3 NP_033904.2 1 - 2 70 72
GenBank mRNA AK088534.1 - 1 - 2 71 72
AK138004.1 - 1 - 2 71 72
BC025089.1 - 1 - 2 82 72
U67327.1 - 1 - 2 55 72

View Full Product Details