Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Kcnk16
- Entrez Gene ID
-
74571
- Gene Name
- potassium channel, subfamily K, member 16
- Gene Aliases
-
4731413G05Rik, TALK-1, TALK1
- Chromosome Location
-
Chr.14: 20262756 - 20269162 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_029006.1 |
NP_083282.1 |
3 - 4 |
499 |
65 |
View Full Product Details