Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp54
Entrez Gene ID
78787
Gene Name
ubiquitin specific peptidase 54
Gene Aliases
4930429G18Rik, AI115571, C030002J06Rik
Chromosome Location
Chr.14: 20548912 - 20643129 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030180.2 NP_084456.2 8 - 9 1560 76
GenBank mRNA AK036864.1 - 7 - 8 949 76
AK047620.1 - 8 - 9 1560 76

View Full Product Details