Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sbf2
Entrez Gene ID
319934
Gene Name
SET binding factor 2
Gene Aliases
4833411B01Rik, AA414977, AI317167, B430219L04Rik, Mtmr13, mMTMH1
Chromosome Location
Chr.7: 110308011 - 110614969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177324.2 NP_796298.2 27 - 28 3802 114
GenBank mRNA BC015069.1 - 4 - 5 593 114
CD538190.1 - 2 - 3 333 114

View Full Product Details