The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031190.2 | NP_112467.2 | 1 - 1 | 1524 | 110 |
GenBank mRNA | AK015016.1 | - | 1 - 1 | 1514 | 110 |
AK133436.1 | - | 1 - 1 | 1513 | 110 | |
BC052343.1 | - | 1 - 1 | 1488 | 110 | |
BC061054.1 | - | 1 - 1 | 1515 | 110 |