Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfyve19
Entrez Gene ID
72008
Gene Name
zinc finger, FYVE domain containing 19
Gene Aliases
1500041L05Rik, ANCHR
Chromosome Location
Chr.2: 119208720 - 119217050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164827.1 NP_001158299.1 3 - 4 640 157
NM_028054.3 NP_082330.2 3 - 4 640 157
GenBank mRNA AK005386.1 - 3 - 4 404 157
BC018420.1 - 3 - 4 335 157
BC099429.1 - 3 - 4 333 157

View Full Product Details