Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfyve19
Entrez Gene ID
72008
Gene Name
zinc finger, FYVE domain containing 19
Gene Aliases
1500041L05Rik, ANCHR
Chromosome Location
Chr.2: 119208720 - 119217050 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164827.1 NP_001158299.1 8 - 9 1292 59
NM_028054.3 NP_082330.2 8 - 9 1292 59
GenBank mRNA AK005386.1 - 8 - 9 1056 59
BC018420.1 - 8 - 9 987 59
BC099429.1 - 8 - 9 985 59
CO038937.1 - 3 - 4 59

View Full Product Details