Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcan2
Entrez Gene ID
53901
Gene Name
regulator of calcineurin 2
Gene Aliases
Csp2, Dscr1l1, MCIP2, ZAKI-4
Chromosome Location
Chr.17: 43801281 - 44039516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207649.1 NP_997532.1 1 - 2 246 97
GenBank mRNA AK136805.1 - 1 - 2 54 97
AK138768.1 - 1 - 2 158 97
BB635249.1 - 1 - 2 158 97
BF534085.1 - - 93 97
BY740155.1 - 1 - 2 253 97

View Full Product Details