Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrg4
Entrez Gene ID
228413
Gene Name
proline rich Gla (G-carboxyglutamic acid) 4 (transmembrane)
Gene Aliases
9930111I18Rik, TMG4
Chromosome Location
Chr.2: 104830741 - 104849850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178695.5 NP_848810.3 5 - 6 783 64
GenBank mRNA AK035530.1 - 5 - 6 608 64
AK037087.1 - 5 - 6 703 64
AK038703.1 - 5 - 6 783 64
AK166405.1 - 4 - 5 744 64
BC099390.1 - 5 - 6 700 64
BC115470.2 - 5 - 6 529 64
BC151011.1 - 5 - 6 713 64
BC151019.1 - 5 - 6 713 64

View Full Product Details