Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mocs1
Entrez Gene ID
56738
Gene Name
molybdenum cofactor synthesis 1
Gene Aliases
3110045D15Rik, AI043170, AW536397
Chromosome Location
Chr.17: 49428364 - 49455435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028464.1 NP_082740.1 10 - 11 1243 100
GenBank mRNA AF214016.1 - 4 - 5 418 100
BC027444.1 - 2 - 3 154 100
BC034066.1 - 6 - 7 592 100
BC049914.1 - 8 - 9 871 100
BC147411.1 - 1 - 2 72 100

View Full Product Details