Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fjx1
Entrez Gene ID
14221
Gene Name
four jointed box 1 (Drosophila)
Gene Aliases
-
Chromosome Location
Chr.2: 102449366 - 102451792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010218.2 NP_034348.2 1 - 1 2034 68
GenBank mRNA BC051990.1 - 1 - 1 2034 68

View Full Product Details