The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172663.4 | NP_766251.3 | 2 - 3 | 629 | 73 |
GenBank mRNA | AK031025.1 | - | 2 - 3 | 602 | 73 |
AK047947.1 | - | 2 - 3 | 297 | 73 | |
BC048785.1 | - | 1 - 2 | 68 | 73 |