Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
H2-Eb2
Entrez Gene ID
381091
Gene Name
histocompatibility 2, class II antigen E beta2
Gene Aliases
A130038H09Rik, H-2Eb2, Ia-5, Ia5, Iebeta2
Chromosome Location
Chr.17: 34325500 - 34341410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033978.3 NP_001029150.1 1 - 2 118 69
GenBank mRNA AK137912.1 - 1 - 2 119 69
AK157199.1 - 1 - 2 129 69
BC150834.1 - 1 - 2 119 69
KJ650234.1 - 1 - 2 101 69

View Full Product Details