Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mtm1
Entrez Gene ID
17772
Gene Name
X-linked myotubular myopathy gene 1
Gene Aliases
AF073996, Mtm, mKIAA4176
Chromosome Location
Chr.X: 71210767 - 71315413 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164190.1 NP_001157662.1 8 - 9 872 66
NM_001164191.1 NP_001157663.1 8 - 9 763 66
NM_001164192.1 NP_001157664.1 8 - 9 763 66
NM_019926.3 NP_064310.2 8 - 9 752 66
GenBank mRNA AF073996.1 - 8 - 9 753 66
AK016445.1 - 8 - 9 873 66
AK031794.1 - 8 - 9 734 66
AK171475.1 - 8 - 9 741 66
BC051022.1 - 8 - 9 721 66
BC090984.1 - 8 - 9 730 66

View Full Product Details