Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atg10
Entrez Gene ID
66795
Gene Name
autophagy related 10
Gene Aliases
5330424L23Rik, 5430428K15Rik, AI852123, APG10, Agp10, Apg10l, Apg10p, Atg10l
Chromosome Location
Chr.13: 90935348 - 91224792 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025770.3 NP_080046.3 4 - 5 403 80
GenBank mRNA AB079383.1 - 3 - 4 359 80
AB091691.1 - 3 - 4 380 80
AK009076.1 - 4 - 5 387 80
AK017355.1 - 4 - 5 385 80
AK077333.1 - 4 - 5 376 80
BC023455.1 - 3 - 4 379 80

View Full Product Details