Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xlr5b
Entrez Gene ID
627081
Gene Name
X-linked lymphocyte-regulated 5B
Gene Aliases
OTTMUSG00000017646
Chromosome Location
Chr.X: 73146976 - 73158886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111293.1 NP_001104763.1 7 - 8 620 108

View Full Product Details