The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153170.3 | NP_694810.2 | 9 - 10 | 1278 | 72 |
GenBank mRNA | AF453744.1 | - | 9 - 10 | 1231 | 72 |
AF512429.1 | - | 9 - 10 | 1301 | 72 | |
AK041041.1 | - | 9 - 10 | 1270 | 72 | |
AK079053.1 | - | 9 - 10 | 1243 | 72 | |
AK156409.1 | - | 9 - 10 | 1279 | 72 | |
AK160394.1 | - | 9 - 10 | 1243 | 72 | |
BC044800.1 | - | 9 - 10 | 1278 | 72 |