Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc36a3
Entrez Gene ID
215332
Gene Name
solute carrier family 36 (proton/amino acid symporter), member 3
Gene Aliases
PAT3, TRAMD2, tramdorin2
Chromosome Location
Chr.11: 55124815 - 55151745 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172258.3 NP_758462.2 6 - 7 766 77
GenBank mRNA AF453745.1 - 6 - 7 766 77
AK132910.1 - 7 - 8 852 77
AY211261.1 - 6 - 7 748 77

View Full Product Details