Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd31
Entrez Gene ID
625662
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
Chromosome Location
Chr.13: 96748289 - 96909794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BG083479.2 - 3 - 4 282 137

View Full Product Details