Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Homer1
Entrez Gene ID
26556
Gene Name
homer homolog 1 (Drosophila)
Gene Aliases
PSD-Zip45, SYN47, Ves-1, homer-1, vesl-1
Chromosome Location
Chr.13: 93304495 - 93405129 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284189.1 NP_001271118.1 2 - 3 956 114
NM_011982.3 NP_036112.1 2 - 3 956 114
NM_147176.3 NP_671705.2 2 - 3 956 114
NM_152134.3 NP_687036.1 2 - 3 475 114
GenBank mRNA AB019478.1 - 2 - 3 390 114
AB019479.1 - 2 - 3 652 114
AB089435.1 - 2 - 3 267 114
AF093257.1 - 2 - 3 958 114
AF093258.1 - 2 - 3 305 114
AK137151.1 - - 1695 114
AY137385.1 - 2 - 3 475 114
BC064041.1 - 2 - 3 776 114

View Full Product Details