Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smn1
Entrez Gene ID
20595
Gene Name
survival motor neuron 1
Gene Aliases
AI849087, Gemin1, Smn
Chromosome Location
Chr.13: 100123205 - 100137699 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252629.1 NP_001239558.1 8 - 9 973 100
NM_011420.2 NP_035550.1 8 - 9 909 100
GenBank mRNA AI849087.1 - 1 - 2 100
AK144778.1 - 8 - 9 910 100
AK152495.1 - 8 - 9 883 100
AK167832.1 - 8 - 9 907 100
AY821787.1 - 5 - 6 278 100
BC037436.1 - 3 - 4 358 100
BC045158.1 - 8 - 9 873 100
EU234021.1 - 8 - 9 973 100
U63294.1 - 8 - 9 887 100
U77714.1 - 8 - 9 870 100
Y12835.1 - 8 - 9 895 100

View Full Product Details