Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
P2ry14
Entrez Gene ID
140795
Gene Name
purinergic receptor P2Y, G-protein coupled, 14
Gene Aliases
A330108O13Rik, Gpr105, P2Y14
Chromosome Location
Chr.3: 59114623 - 59153492 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008497.2 NP_001008497.1 1 - 2 267 137
NM_133200.4 NP_573463.1 1 - 2 267 137
GenBank mRNA AF177211.1 - 1 - 2 128 137
AK140301.1 - 1 - 2 99 137
BC058558.1 - 1 - 2 129 137

View Full Product Details