Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mis18bp1
Entrez Gene ID
217653
Gene Name
MIS18 binding protein 1
Gene Aliases
6720403H11, C79407, Knl2, mKIAA1903
Chromosome Location
Chr.12: 65132734 - 65172580 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172578.2 NP_766166.2 3 - 4 1232 135
GenBank mRNA AK032698.1 - 1 - 2 263 135
AK035117.1 - 3 - 4 1234 132
AK053496.1 - 3 - 4 1242 135
AK162657.1 - 3 - 4 1256 135
AK220223.1 - 2 - 3 498 135
BC042728.1 - 3 - 4 1250 135
BC052175.1 - 3 - 4 1232 135

View Full Product Details