Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pde8b
Entrez Gene ID
218461
Gene Name
phosphodiesterase 8B
Gene Aliases
B230331L10Rik, C030047E14Rik
Chromosome Location
Chr.13: 95024105 - 95250397 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170669.1 NP_001164140.1 6 - 7 1032 124
NM_172263.2 NP_758467.2 6 - 7 624 124
GenBank mRNA AK036090.1 - 7 - 8 952 124
AK043680.1 - 6 - 7 1025 124
AK045991.1 - 6 - 7 1033 124
AK046298.1 - 6 - 7 1048 124
AK083166.1 - 6 - 7 670 124
BC032286.1 - 6 - 7 636 124
EF644173.1 - 6 - 7 486 124

View Full Product Details