Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr41
Entrez Gene ID
218460
Gene Name
WD repeat domain 41
Gene Aliases
B830029I03Rik, MSTP048
Chromosome Location
Chr.13: 94976213 - 95023316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172590.3 NP_766178.2 12 - 13 1288 98
GenBank mRNA AK046853.1 - 12 - 13 1257 98
AK149993.1 - 12 - 13 1254 98
AK194357.1 - 3 - 4 328 98

View Full Product Details