Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr41
Entrez Gene ID
218460
Gene Name
WD repeat domain 41
Gene Aliases
B830029I03Rik, MSTP048
Chromosome Location
Chr.13: 94976213 - 95023316 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172590.3 NP_766178.2 6 - 7 612 63
GenBank mRNA AK082348.1 - 6 - 7 613 63
AK149993.1 - 6 - 7 578 63
BB650073.1 - 6 - 7 608 63
BC138095.1 - 6 - 7 518 63
BC145143.1 - 6 - 7 514 63
BC145144.1 - 6 - 7 518 63

View Full Product Details