The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290500.1 | NP_001277429.1 | 4 - 5 | 661 | 102 |
NM_001290502.1 | NP_001277431.1 | 4 - 5 | 899 | 102 | |
NM_025978.4 | NP_080254.2 | 4 - 5 | 661 | 102 | |
NM_027619.4 | NP_081895.2 | 4 - 5 | 661 | 102 | |
GenBank mRNA | AK012246.1 | - | 4 - 5 | 623 | 102 |
AK015308.1 | - | 4 - 5 | 621 | 102 | |
AK017763.1 | - | 2 - 3 | 259 | 102 | |
AK031605.1 | - | 4 - 5 | 892 | 102 | |
AK035570.1 | - | 4 - 5 | 631 | 102 | |
AK041095.1 | - | 4 - 5 | 635 | 102 | |
AK044266.1 | - | 4 - 5 | 636 | 102 | |
AK134539.1 | - | 4 - 5 | 650 | 102 | |
AK173315.1 | - | 4 - 5 | 631 | 102 | |
BC021448.1 | - | 4 - 5 | 669 | 102 | |
BC024847.1 | - | 4 - 5 | 626 | 102 | |
BC138212.1 | - | 4 - 5 | 623 | 102 | |
BC145944.1 | - | 4 - 5 | 623 | 102 |