The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040399.1 | NP_001035489.1 | 3 - 4 | 167 | 126 |
GenBank mRNA | AK160155.1 | - | 3 - 4 | 167 | 126 |
AK163348.1 | - | 3 - 4 | 274 | 126 | |
BC025528.1 | - | 1 - 2 | 90 | 126 |