Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Intu
Entrez Gene ID
380614
Gene Name
inturned planar cell polarity effector homolog (Drosophila)
Gene Aliases
9230116I04Rik, 9430087H23Rik, Pdzd6, Pdzk6, mKIAA1284
Chromosome Location
Chr.3: 40653702 - 40704774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175515.5 NP_780724.3 - 2361 71
GenBank mRNA AK035092.1 - - 2342 71
AK122485.1 - 11 - 12 1901 71
BC125523.1 - - 2308 71
BC132387.1 - - 2308 71

View Full Product Details