Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo4
Entrez Gene ID
226564
Gene Name
flavin containing monooxygenase 4
Gene Aliases
D1Ertd532e
Chromosome Location
Chr.1: 162793188 - 162815869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK146562.1 - 2 - 3 227 65

View Full Product Details