Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo6
Entrez Gene ID
226565
Gene Name
flavin containing monooxygenase 6
Gene Aliases
-
Chromosome Location
Chr.1: 162916551 - 162937572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178038.1 NP_001171509.1 3 - 4 584 70
GenBank mRNA BC051402.1 - 4 - 5 671 70

View Full Product Details