The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164059.1 | NP_001157531.1 | 7 - 8 | 1100 | 101 |
NM_011346.2 | NP_035476.1 | 8 - 9 | 1208 | 101 | |
GenBank mRNA | AK137509.1 | - | 8 - 9 | 1120 | 101 |
AK157273.1 | - | 8 - 9 | 1197 | 101 | |
AK161754.1 | - | 8 - 9 | 1200 | 101 | |
AK163066.1 | - | 6 - 7 | 665 | 101 | |
AK170753.1 | - | 7 - 8 | 1101 | 101 | |
AK172116.1 | - | 8 - 9 | 1198 | 101 | |
BC052681.1 | - | 8 - 9 | 1192 | 101 | |
M25324.1 | - | 8 - 9 | 1200 | 101 | |
M36005.1 | - | 8 - 9 | 1146 | 101 | |
M36058.1 | - | 8 - 9 | 1146 | 101 | |
X14772.1 | - | 8 - 9 | 1149 | 101 |