Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Baz1a
Entrez Gene ID
217578
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
Acf1, B930060C03, BC065123, Gtl5, Wcrf180, cbp146
Chromosome Location
Chr.12: 54892600 - 55014371 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013815.2 NP_038843.2 7 - 8 1063 92
GenBank mRNA AF033664.1 - 6 - 7 808 92
AK135668.1 - 7 - 8 1063 92
AK166955.1 - 4 - 5 382 92

View Full Product Details