Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trmt1l
Entrez Gene ID
98685
Gene Name
tRNA methyltransferase 1 like
Gene Aliases
1190005F20Rik, AW226554, C1orf25, Trm1-like, Trm1l
Chromosome Location
Chr.1: 151428547 - 151458183 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026876.3 NP_081152.2 8 - 9 1328 78
GenBank mRNA AK146088.1 - 8 - 9 1329 78
BC022682.1 - 3 - 4 364 78
BC037478.1 - 4 - 5 508 78
BC061491.1 - 8 - 9 1334 78
BC064024.1 - 8 - 9 1126 78
BC110425.1 - 8 - 9 1311 78
BC132299.1 - 8 - 9 1186 78

View Full Product Details