Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Allc
Entrez Gene ID
94041
Gene Name
allantoicase
Gene Aliases
1700012B22Rik, Alc
Chromosome Location
Chr.12: 28553755 - 28582552 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053156.2 NP_444386.2 7 - 8 704 68
GenBank mRNA AF278712.1 - 7 - 8 712 68
BC108957.1 - 7 - 8 704 68
BC108958.1 - 7 - 8 704 68

View Full Product Details