Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mme
Entrez Gene ID
17380
Gene Name
membrane metallo endopeptidase
Gene Aliases
6030454K05Rik, C85356, CALLA, CD10, NEP, SFE
Chromosome Location
Chr.3: 63295226 - 63383713 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289462.1 NP_001276391.1 4 - 5 809 113
NM_001289463.1 NP_001276392.1 4 - 5 593 113
NM_008604.4 NP_032630.2 4 - 5 443 113
GenBank mRNA AK031446.1 - 4 - 5 444 113
AK033824.1 - 4 - 5 504 113
AK040730.1 - 4 - 5 480 113
AK052088.1 - 3 - 4 330 113
AK076908.1 - 4 - 5 657 113
AK080202.1 - 4 - 5 471 113
BC034092.1 - 4 - 5 465 113
M81591.1 - 3 - 4 355 113

View Full Product Details