Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lct
Entrez Gene ID
226413
Gene Name
lactase
Gene Aliases
Gm100, LAC, LPH, Lphl
Chromosome Location
Chr.1: 128284764 - 128328318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081078.1 NP_001074547.1 15 - 16 5363 70
GenBank mRNA AK158042.1 - 9 - 10 3465 70
AY751548.1 - 3 - 4 464 70

View Full Product Details