The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081011.2 | NP_001074480.2 | 20 - 21 | 3080 | 113 |
GenBank mRNA | AK132220.1 | - | 6 - 7 | 753 | 113 |
AK161932.1 | - | 9 - 10 | 984 | 113 | |
AY057900.1 | - | 11 - 12 | 1069 | 113 | |
BC030457.1 | - | 19 - 20 | 2387 | 113 | |
BC151081.1 | - | 19 - 20 | 2698 | 113 | |
BC151082.1 | - | 19 - 20 | 2698 | 113 | |
BC158055.1 | - | 19 - 20 | 2696 | 113 | |
BC172152.1 | - | 19 - 20 | 2698 | 113 |