The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024186.5 | NP_077148.1 | 5 - 6 | 433 | 157 |
NM_024272.4 | NP_077234.2 | 6 - 7 | 523 | 247 | |
GenBank mRNA | AK010220.1 | - | 5 - 6 | 375 | 157 |
AK013177.1 | - | 6 - 7 | 449 | 247 | |
AK051840.1 | - | 5 - 6 | 370 | 157 | |
AK133699.1 | - | 5 - 6 | 377 | 157 | |
AY037837.1 | - | 6 - 7 | 479 | 247 | |
AY167988.1 | - | 6 - 7 | 433 | 247 | |
BC002176.1 | - | 5 - 6 | 445 | 157 |