Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfyve16
Entrez Gene ID
218441
Gene Name
zinc finger, FYVE domain containing 16
Gene Aliases
AI035632, B130024H06Rik, B130031L15, mKIAA0305
Chromosome Location
Chr.13: 92487104 - 92532710 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173392.4 NP_775568.1 1 - 2 106 99
GenBank mRNA AK045090.1 - 1 - 2 164 99
AK122241.1 - 1 - 2 87 99
AK170435.1 - 1 - 2 125 99
BC042669.1 - 1 - 2 104 99

View Full Product Details